Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XNDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001374469.1 | NP_001361398.1 | 5 - 6 | 926 | 99 | |
GenBank mRNA | BC150095.1 | - | 5 - 6 | 917 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |