Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099372.1 | NP_001092842.1 | 6 - 7 | 510 | 74 | |
GenBank mRNA | BC146107.1 | - | 6 - 7 | 510 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |