Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100371.1 | NP_001093841.1 | 6 - 7 | 1990 | 92 | |
GenBank mRNA | BC149232.1 | - | 6 - 7 | 1990 | 92 | |
BT026149.1 | - | 2 - 3 | 794 | 92 | ||
BT026332.1 | - | 2 - 3 | 754 | 92 | ||
BT029787.1 | - | 2 - 3 | 799 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |