Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL23R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127172.1 | NP_001120644.1 | 6 - 7 | 958 | 111 | |
GenBank mRNA | EU616678.1 | - | 6 - 7 | 958 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |