Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPTL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128070.1 | NP_001121542.1 | 1 - 2 | 612 | 83 | |
GenBank mRNA | EU599229.1 | - | 1 - 2 | 612 | 83 | |
JN848788.1 | - | 1 - 2 | 579 | 83 | ||
JN848789.1 | - | 1 - 2 | 579 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Bos taurus Probe spans exons |