Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130747.1 | NP_001124219.1 | 4 - 5 | 961 | 74 | |
GenBank mRNA | BC118284.1 | - | 4 - 5 | 961 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |