Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166515.1 | NP_001159987.1 | 4 - 5 | 1706 | 100 | |
GenBank mRNA | AF192771.1 | - | 3 - 4 | 488 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |