Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULT2B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081726.1 | NP_001075195.1 | 4 - 5 | 590 | 77 | |
GenBank mRNA | BC126756.1 | - | 4 - 5 | 590 | 77 | |
DQ114541.1 | - | 2 - 3 | 144 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |