Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP26A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101185.1 | NP_001094655.1 | 6 - 7 | 1187 | 93 | |
NM_001168662.1 | NP_001162133.1 | 5 - 6 | 989 | 93 | ||
GenBank mRNA | BC149355.1 | - | 5 - 6 | 990 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |