Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101881.2 | NP_001095351.2 | 6 - 7 | 948 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |