Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RETSAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102279.2 | NP_001095749.2 | 10 - 11 | 1732 | 67 | |
GenBank mRNA | BC149535.1 | - | 11 - 12 | 1678 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |