Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102553.1 | NP_001096023.1 | 2 - 3 | 150 | 83 | |
GenBank mRNA | BC114697.1 | - | 2 - 3 | 150 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |