Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105325.2 | NP_001098795.1 | 3 - 4 | 286 | 106 | |
GenBank mRNA | AF419316.1 | - | 2 - 3 | 116 | 106 | |
BC102100.1 | - | 3 - 4 | 287 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |