Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HES5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105467.1 | NP_001098937.1 | 1 - 2 | 113 | 62 | |
GenBank mRNA | BC146153.1 | - | 1 - 2 | 113 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |