Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B9D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110064.2 | NP_001103534.2 | 1 - 2 | 100 | 92 | |
GenBank mRNA | BC109549.1 | - | 1 - 2 | 102 | 92 | |
EV674790.1 | - | 1 - 2 | 86 | 92 | ||
EW682825.1 | - | 1 - 2 | 89 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |