Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161651.1 | NP_001155123.1 | 18 - 19 | 2214 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Bos taurus Probe spans exons |