Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELENOO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163193.3 | NP_001156665.2 | 5 - 6 | 1381 | 60 | |
GenBank mRNA | DT814669.1 | - | 2 - 3 | 358 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |