Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIAH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205421.2 | NP_001192350.2 | 1 - 2 | 138 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Bos taurus Probe spans exons |