Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL17RD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205427.2 | NP_001192356.2 | 3 - 4 | 315 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |