Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETD1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205433.3 | NP_001192362.3 | 1 - 2 | 146 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |