Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCGB3A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205460.2 | NP_001192389.1 | 1 - 2 | 159 | 61 | |
GenBank mRNA | EV633292.1 | - | 1 - 2 | 162 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Bos taurus Probe spans exons |