Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205470.1 | NP_001192399.1 | 18 - 19 | 1813 | 84 | |
GenBank mRNA | BC151667.1 | - | 18 - 19 | 1785 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |