Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LNP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205535.2 | NP_001192464.1 | 1 - 2 | 310 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |