Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDFI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205537.1 | NP_001192466.1 | 3 - 4 | 475 | 87 | |
GenBank mRNA | AF184162.1 | - | 1 - 2 | 44 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |