Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5C1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205546.3 | NP_001192475.1 | 4 - 5 | 621 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |