Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205547.3 | NP_001192476.2 | 1 - 2 | 229 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |