Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205561.1 | NP_001192490.1 | 4 - 5 | 409 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |