Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WTIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205562.1 | NP_001192491.1 | 3 - 4 | 839 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |