Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THRAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205565.1 | NP_001192494.1 | 10 - 11 | 2693 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |