Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205583.1 | NP_001192512.1 | 5 - 6 | 507 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Bos taurus Probe spans exons |