Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKAA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205605.1 | NP_001192534.1 | 6 - 7 | 784 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |