Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC4G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205606.3 | NP_001192535.3 | 2 - 3 | 202 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |