Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205649.1 | NP_001192578.1 | 5 - 6 | 1389 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Bos taurus Probe spans exons |