Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205657.2 | NP_001192586.2 | 4 - 5 | 563 | 120 | |
GenBank mRNA | CV981480.1 | - | 4 - 5 | 464 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |