Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205658.1 | NP_001192587.1 | 7 - 8 | 1735 | 82 | |
GenBank mRNA | BC149001.1 | - | 8 - 9 | 2292 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |