Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205669.1 | NP_001192598.1 | 6 - 7 | 982 | 58 | |
GenBank mRNA | BT025405.1 | - | 6 - 7 | 981 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |