Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205672.1 | NP_001192601.1 | 10 - 11 | 1224 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |