Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205679.2 | NP_001192608.2 | 4 - 5 | 638 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |