Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OIP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205692.1 | NP_001192621.1 | 4 - 5 | 629 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |