Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205721.1 | NP_001192650.1 | 3 - 4 | 598 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |