Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205751.2 | NP_001192680.2 | 3 - 4 | 1358 | 57 | |
GenBank mRNA | BC148920.1 | - | 3 - 4 | 1090 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |