Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205761.3 | NP_001192690.1 | 8 - 9 | 1464 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Bos taurus Probe spans exons |