Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMND5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205832.1 | NP_001192761.1 | 8 - 9 | 1108 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Bos taurus Probe spans exons |