Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205833.3 | NP_001192762.2 | 4 - 5 | 486 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons |