Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205835.1 | NP_001192764.1 | 3 - 4 | 764 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |