Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205849.3 | NP_001192778.3 | 14 - 15 | 1865 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |