Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNNM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205896.2 | NP_001192825.2 | 1 - 2 | 1409 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |