Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205899.3 | NP_001192828.1 | 14 - 15 | 2014 | 111 | |
NM_001317187.2 | NP_001304116.1 | 14 - 15 | 2014 | 111 | ||
GenBank mRNA | EE347213.1 | - | 6 - 7 | 770 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |