Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205899.3 | NP_001192828.1 | 2 - 3 | 385 | 103 | |
NM_001317187.2 | NP_001304116.1 | 2 - 3 | 385 | 103 | ||
GenBank mRNA | EE913054.1 | - | 2 - 3 | 385 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |