Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205938.1 | NP_001192867.1 | 11 - 12 | 2015 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |